Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Control: mouse brain, rat brain
Predicted reactivity: Rat (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RASGRF1 is a guanine nucleotide exchange factor (GEF) similar to the Saccharomyces cerevisiae CDC25 gene product. Functional analysis has demonstrated that this protein stimulates the dissociation of GDP from RAS protein. Studies of the similar gene in mouse suggested that the Ras-GEF activity of this protein in brain can be activated by Ca2+ influx, muscarinic receptors, and G protein beta-gamma subunit. Mouse studies also indicated that the Ras-GEF signaling pathway mediated by this protein may be important for long-term memory.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASPP1; CDC25Mm; GNRP; Guanine nucleotide-releasing protein; KIAA0771; P140 RAS-GRF; PPP1R13B; Ras guanine release factor beta; Ras-GRF1; Ras-specific guanine nucleotide-releasing factor 1; Ras-specific nucleotide exchange factor CDC25
Gene Aliases: AI844718; CDC25; CDC25Mm; Gnrp; Grf1; Grfbeta; p190; P190-A; p190RhoGEF; Ras-GRF1; Rasgrf1
UniProt ID: (Mouse) P27671, (Rat) P28818
Entrez Gene ID: (Mouse) 19417, (Rat) 192213
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support