Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SLQNASHHLE PNMDLINLLT LSLDLYHTED DIYKLSLVLE PRNSKSQPTS PTTPNKPVVP LEWALGVMPK PDPTVINKHI RKLVESVFRN YDHDHDGYIS QEDFESIAAN FPFLDSFCVL DKDQDGLISK DEMMAYFLRA KSQLHCKMGP GFIHNFQEMT YLKPTFCEHC AGFLWGIIKQ GYKCKDCGAN CHKQCKDLLV LACRRFARAP SLSSGHGSLP GSPSLPPAQD EVFEFPGVTA GHRDLDSRAI TLVTGSSRKI SVRLQRATTS QATQTEPVWS EAGWGDSGSH TFPKMKSKFH DKAAKDKGFA KWENEKPRVH AGVDVVDRGT EFELDQDEGE ETRQDGEDG
Members of the RAS subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RASGRP3, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium and DAG-regulated guanine nucleotide exchange factor III; calcium- and diacylglycerol-regulated guanine nucleotide exchange factor III; CalDAG-GEFIII; Guanine nucleotide exchange factor for Rap1; OTTHUMP00000201245; RAS guanyl releasing protein 3 (calcium and DAG-regulated); Ras guanyl-releasing protein 3
Gene Aliases: BC066069; Gm327; GRP3; KIAA0846; RASGRP3
UniProt ID: (Human) Q8IV61
Entrez Gene ID: (Human) 25780, (Mouse) 240168
Molecular Function: guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support