Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MADKEAAFDD AVEERVINEE YKIWKKNTPF LYDLVMTHAL EWPSLTAQWL PDVTRPEGKD FSIHRLVLGT HTSDEQNHLV IASVQLPNDD AQFDASHYDS EKGEFGGFGS VSGKIEIEIK INHEGEVNRA RYMPQNPCII ATKTPSSDVL VFDYTKHPSK PDPSGECNPD LRLRGHQKEG YGLSWNPNLS GHLLSASDDH TICLWDISAV PKEGKVVDAK TIFTGHTAVV EDVSWHLLHE SLFGSVADDQ KLMIWDTRSN NTSKPSHSVD AHTAEVNCLS FNPYSEFILA TGSADKTVAL WDLRNLKLKL HSFESHKDEI FQVQWSPHNE TILASSGTDR RLNVWDLSKI GEEQSPEDAE DGPPELLFIH GGHTAKISDF SWNPNEPWVI CSVSEDNIMQ VWQMAENIYN DEDPEGSVDP EGQGS
RbAp48-11G10 recognizes full length RbAp48, a 48 kDa nuclear protein first isolated by its ability to interact with the carboxyl-terminus of the retinoblastoma protein (Rb). It is a member of the WD (Trp-Asp) repeat family of proteins. RbAp48 is one of the three subunits of CAF-1, can bind to histone H4 and is a subunit of human histone deacetylase HD1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAF-1 p48 subunit; CAF-1 subunit C; CAF-I 48 kDa subunit; CAF-I p48; Chromatin assembly factor 1 subunit C; Chromatin assembly factor I p48 subunit; chromatin assembly factor/CAF-1 p48 subunit; Histone-binding protein RBBP4; MSI1 protein homolog; Nucleosome-remodeling factor subunit RBAP48; RBBP-4; retinoblastoma binding protein 4; Retinoblastoma-binding protein 4; Retinoblastoma-binding protein p48
Gene Aliases: lin-53; mRbAp48; NURF55; RBAP48; RBBP4
UniProt ID: (Human) Q09028, (Mouse) Q60972
Entrez Gene ID: (Human) 5928, (Rat) 313048, (Mouse) 19646
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support