Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide Sequence: MAASGEPQRQ WQEEVAAVVV VGSCMTDLVS LTSRLPKTGE TIHGHKFFIG FGGKGANQCV QAARLGAMTS MVCKVGKDSF GNDYIENLKQ NDISTEFTYQ TKDAATGTAS IIVNNEGQNI IVIVAGANLL LNTEDLRAAA NVISRAKVMV CQLEITPATS LEALTMARRS GVKTLFNPAP AIADLDPQFY TLSDVFCCNE SEAEILTGLT VGSAADAGEA ALVLLKRGCQ VVIITLGAEG CVVLSQTEPE PKHIPTEKVK AVDTTGAGDS FVGALAFYLA YYPNLSLEDM LNRSNFIAAV SVQAAGTQSS YPYKKDLPLT LF
RBSK belongs to the pfkB family of carbohydrate kinases. It phosphorylates ribose to form ribose-5-phosphate in the presence of ATP and magnesium as a first step in ribose metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5230400M11Rik; Ribokinase; RK
Gene Aliases: RBKS; RBSK; RK
UniProt ID: (Human) Q9H477
Entrez Gene ID: (Human) 64080
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support