Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Aliquoting is unnecessary for -20°C storage.
May function in the regulation of alternative or illicit splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromosome 3p21.1 gene sequence; HsOTT3; huOTT3; one twenty two protein 3; One-twenty two protein 3; Putative RNA-binding protein 15B; rna-binding; RNA-binding motif protein 15B
Gene Aliases: HUMAGCGB; OTT3; RBM15B
UniProt ID: (Human) Q8NDT2
Entrez Gene ID: (Human) 29890
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support