Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AKAEETRYPQ QYQPSPLPVH YELLTDGYTR HRNLDADLVR DRTPPHLLYS DRDRTFLEGD WTSPSKSSDR RNSLEGYSRS VRSRSGERWG ADGDRGLPKP WEERRKRRSL SSDRGRTTHS PYEERSRTKG SGQQSERGSD RTPERSRKEN HSSEGTKESS SNSLSNSRHG AEERGHHHHH HEAADSSHGK KARDSERNHR TTEAEPKPLE EPKHETKKLK NLSEYAQTLQ LGWNGLLVLK NSCFPTSMHI LEGDQGVISS LLKDHTSGSK LTQLKIAQRL
May function in the regulation of alternative or illicit splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromosome 3p21.1 gene sequence; HsOTT3; huOTT3; one twenty two protein 3; One-twenty two protein 3; Putative RNA-binding protein 15B; rna-binding; RNA-binding motif protein 15B
Gene Aliases: HUMAGCGB; OTT3; RBM15B
UniProt ID: (Human) Q8NDT2
Entrez Gene ID: (Human) 29890
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support