Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSEEGKLFV GGLNFNTDEQ ALEDHFSSFG PISEVVVVKD RETQRSRGFG FITFTNPEHA SVAMRAMNGE SLDGRQIRVD
RBM3 is a member of the glycine-rich RNA-binding protein family and a protein with one RNA recognition motif (RRM) domain. Expression of this protein is induced by cold shock and low oxygen tension.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OTTHUMP00000025800; OTTHUMP00000025802; putative RNA-binding protein 3; RNA-binding motif protein 3; RNA-binding protein 3; RNPL
Gene Aliases: IS1-RNPL; RBM3; RNPL
UniProt ID: (Human) P98179
Entrez Gene ID: (Human) 5935
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support