Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: HeLa, HeLa nucleus.
Predicted reactivity: Human (99%), Mouse (85%), Rat (88%), Rhesus Monkey (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May bind RNA.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA-binding motif protein 41; RNA-binding protein 41
Gene Aliases: RBM41
UniProt ID: (Human) Q96IZ5
Entrez Gene ID: (Human) 55285
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support