Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C CRABP-I; cellular retinol binding protein 1; Cellular retinol-binding protein; Cellular retinol-binding protein I; CRBP; CRBP-I; RBP; retinol; retinol binding; Retinol Binding Protein; Retinol-binding protein 1; retinol-binding protein 1, cellular
Gene Aliases: CRABP-I; CRBP; CRBP1; CRBPI; RBP1; RBPC
UniProt ID: (Human) P09455
Entrez Gene ID: (Human) 5947
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support