Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKWVWALFL LAALGSGRAE RDCRVSSFRV KENFDKARFS GTWYAMAKKD PEGLFLQDNI VAEFSVDETG QMSATAKGRV RLLNNWDVCA DMVGTFTDTE DPAKFKMKYW GVASFLQKGN DDHWIVDTDY DTYAVQYSCR LLNLDGTCAD SYSFVFSRDP NGLPPEAQKI VRQRQEELCL ARQYRLIVHN GYCDGRSERN LL (1-201 aa encoded by BC020633)
This protein belongs to the lipocalin family and is the specific carrier for retinol in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein post translationally and results in defective delivery and supply to the epidermal cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Plasma retinol binding protein 4 (PRBP); Plasma retinol-binding protein; PRBP; RBP; retinol; retinol binding; Retinol binding protein 4 interstitial; retinol binding protein 4, plasma; Retinol-binding protein 4; retinol-binding protein 4, interstitial
Gene Aliases: MCOPCB10; PRO2222; RBP4; RDCCAS
UniProt ID: (Human) P02753
Entrez Gene ID: (Human) 5950
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support