Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: PTVRKERVVR PDSELGERPP EDNQSFQYDH EAFLGKEDSK TFDQLTPDES KERLGKIVDR IDNDGDGFVT TEELKTWIKR VQKRYIFDNV AKVWKDYDRD KDDKISWEEY KQATYGYYLG NPAEFHDSSD HHTFKKMLPR DERRFKAADL NGDLTATREE FTAFLHPEEF EHMKEIVVLE TLEDIDKNGD GFVDQDEYIA DMFSHEENGP EPDWVLSERE QFNEFRDLNK DGKLDKDEIR HWILPQDYDH AQAEARHLVY ESDKNKDEKL TKEEILENWN MFVGSQATNY GEDLTKNHDE L
The RCAN1 protein interacts with calcineurin A and inhibits calcineurin-dependent signaling pathways, possibly affecting central nervous system development. This gene is located in the minimal candidate region for the Down syndrome phenotype, and is overexpressed in the brain of Down syndrome fetuses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adapt78; Calcipressin-1; calcium and oxidant-inducible mRNA; Down syndrome candidate region 1; Down syndrome critical region gene 1; Down syndrome critical region protein 1; MCIP1; modulatory calcineurin-interacting protein 1; Myocyte-enriched calcineurin-interacting protein 1; near DSCR proline-rich protein; Regulator of calcineurin 1
Gene Aliases: ADAPT78; CSP1; DSC1; DSCR1; MCIP1; RCAN1; RCN1
UniProt ID: (Human) P53805
Entrez Gene ID: (Human) 1827
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support