Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LFRTYDDCV TFQLFKSFRR VRINFSNPKS AARARIELHE TQFRGKKLKL YFAQVQTPET DGDKLHLAPP QPAKQFLISP PSSPPVGWQP INDATPVLNY DLLYAVAKLG PGEKYELHAG TESTPSVVVH VCDSDIEEEE DPKTSPKPKI IQTRRPGLPP SVSNWAAC (35-201 aa encoded by BC038509 )
Regulator of calcineurin 2 (RCAN2), also known as ZAKI4 and DSCR1L1, is expressed as two isoforms differing at their N-terminus. The longer of the two (isoform 1) is expressed exclusively in the brain, while isoform 2 is ubiquitously expressed, with highest expression in brain, heart, and muscle. Both isoforms bind to the catalytic subunit of calcineurin, a Ca++-dependent protein phosphatase involved in several neuronal functions, though their C-terminal region and inhibit calcineurin's activity. Unlike isoform 1 of RCAN2, the expression of the second isoform is not induced by the thyroid hormone T3. RCAN2 is a member of a family of three endogenous calcineurin regulators that are located near the minimal supernumerary fragment of chromosome 21 in individuals with Down syndrome, suggesting that they play a role in this syndrome. Multiple isoforms of RCAN2 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcineurin inhibitory protein ZAKI-4; Calcipressin-2; Down syndrome candidate region 1-like 1; Down syndrome candidate region 1-like protein 1; Down syndrome critical region gene 1-like 1; MCIP2; myocyte-enriched calcineurin interactin protein 2; Myocyte-enriched calcineurin-interacting protein 2; Regulator of calcineurin 2; thyroid hormone-responsive (skin fibroblasts); Thyroid hormone-responsive protein ZAKI-4
Gene Aliases: CSP2; DSCR1L1; MCIP2; RCAN2; RCN2; ZAKI-4; ZAKI4
UniProt ID: (Human) Q14206, (Mouse) Q9JHG2
Entrez Gene ID: (Human) 10231, (Mouse) 53901
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support