Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEEELPLFSG DSGKPVQATL SSLKMLDVGK WPIFSLCSEE ELQLIRQACV FGSAGNEVLY TTVNDEIFVL GTNCCGCLGL GDVQSTIEPR RLDSLNGKKI ACLSYGSGPH IVLATTEGEV FTWGHNAYSQ LGNGTTNHGL VPCHISTNLS NKQVIEVACG SYHSLVLTSD GEVFAWGYNN SGQVGSGSTV NQPIPRRVTG CLQNKVVVTI ACGQMCCMAV VDTGEVYVWG YNGNGQLGLG NSGNQPTPCR VAALQGIRVQ RVACGYAHTL VLTDEGQVYA WGANSYGQLG TGNKSNQSYP TPVTVEKDRI IEIAACHSTH TSAAKTQGGH VYMWGQCRGQ SVILPHLTHF SCTDDVFACF ATPAVTWRLL SVEPDDHLTV AESLKREFDN PDTADLKFLV DGKYIYAHKV LLKIRCEHFR SSLEDNEDDI VEMSEFSYPV YRAFLEYLYT DSISLSPEEA VGLLDLATFY RENRLKKLCQ QTIKQGICEE NAIALLSAAV KYDAQDLEEF CFRFCINHLT VVTQTSGFAE MDHDLLKNFI SKASRVGAFK N
RCBTB2 is a member of the RCC1-related GEF family. The N-terminal half of the amino acid sequence shows similarity to the regulator of chromosome condensation RCC1, which acts as a guanine nucleotide exchange factor (GEF) protein for the Ras-related GTPase Ran.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CHC1-L; Chromosome condensation 1-like; RCC1 and BTB domain-containing protein 2; RCC1-like G exchanging factor; RCC1-like G exchanging factor RLG; regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2; Regulator of chromosome condensation and BTB domain-containing protein 2
Gene Aliases: CHC1L; RCBTB2; RLG
UniProt ID: (Human) O95199
Entrez Gene ID: (Human) 1102
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support