Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is a rabbit polyclonal antibody raised against full length RCC1 of human origin.
Immunogen sequence: MSPKRIAKR RSPPADAIPK SKKVKVSHRS HSTEPGLVLT LGQGDVGQLG LGENVMERKK PALVSIPEDV VQAEAGGMHT VCLSKSGQVY SFGCNDEGAL GRDTSVEGSE MVPGKVELQE KVVQVSAGDS HTAALTDDGR VFLWGSFRDN NGVIGLLEPM KKSMVPVQVQ LDVPVVKVAS GNDHLVMLTA DGDLYTLGCG EQGQLGRVPE LFANRGGRQG LERLLVPKCV MLKSRGSRGH VRFQDAFCGA YFTFAISHEG HVYGFGLSNY HQLGTPGTES CFIPQNLTSF KNSTKSWVGF SGGQHHTVCM DSEGKAYSLG RAEYGRLGLG EGAEEKSIPT LISRLPAVSS VACGASVGYA VTKDGRVFAW GMGTNYQLGT GQDEDAWSPV EMMGKQLENR VVLSVSSGGQ HTVLLVKDKE QS (1-421 aa encoded by BC010067)
Guanine-nucleotide releasing factor that promotes the exchange of Ran-bound GDP by GTP. Involved in the regulation of onset of chromosome condensation in the S phase. Binds both to the nucleosomes and double-stranded DNA. RCC1-Ran complex (together with other proteins) acts as a component of a signal transmission pathway that detects unreplicated DNA. Plays a key role in nucleo-cytoplasmic transport, mitosis and nuclear-envelope assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell cycle regulatory protein; Chromosome condensation protein 1; guanine nucleotide-releasing protein; Regulator of chromosome condensation; SNHG3-RCC1 readthrough
Gene Aliases: 4931417M11Rik; AI326872; CHC1; RCC1; RCC1-I; SNHG3-RCC1
UniProt ID: (Human) P18754, (Mouse) Q8VE37
Entrez Gene ID: (Human) 1104, (Mouse) 100088, (Rat) 682908
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support