Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QLSMDCPCDL ADGLKVVLAP RSWARCLTDM RWLRNQVIAP LTEELVFRAC
This gene encodes an integral membrane protein which is classified as a member of the metalloproteinase family. This enzyme is thought to function in the maintenance and processing of CAAX-type prenylated proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAAX prenyl protease 2; FACE-2; farnesylated protein-converting enzyme 2; Farnesylated proteins-converting enzyme 2; hRCE1; Prenyl protein-specific endoprotease 2; RCE1 homolog; RCE1 homolog, prenyl protein peptidase
Gene Aliases: FACE2; RCE1; RCE1A; RCE1B
UniProt ID: (Human) Q9Y256
Entrez Gene ID: (Human) 9986
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support