Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MATQAHSLSY AGCNFLRQRL VLSTLSGRPV KIRKIRARDD NPGLRDFEAS FIRLLDKITN GSRIEINQTG TTLYYQPGLL YGGSVEHDCS VLRGIGYYLE SLLCLAPFMK HPLKIVLRGV TNDQVDPSVD VLKATALPLL KQFGIDGESF ELKIVRRGMP PGGGGEVVFS CPVRKVLKPI QLTDPGKIKR IRGMAYSVRV SPQMANRIVD SARSILNKFI PDIYIYTDHM KGVNSGKSPG FGLSLVAETT SGTFLSAELA SNPQGQGAAV LPEDLGRNCA RLLLEEIYRG GCVDSTNQSL ALLLMTLGQQ DVSKVLLGPL SPYTIEFLRH LKSFFQIMFK IETKPCGEEL KGGDKVLMTC VGIGFSNLSK TLK
RCL1, which does not have cyclase activity, plays a role in 40S- ribosomal-subunit biogenesis in the early pre-rRNA processing steps at sites A0, A1 and A2 that are required for proper maturation of the 18S RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA 3'-terminal phosphate cyclase-like protein; RNA cyclase homolog; RNA terminal phosphate cyclase-like 1
Gene Aliases: 2310040A02Rik; Ab1-353; Ac1292; AI789745; C76567; HSPC338; RCL1; RNAC; RPC2; RPCL1; RTC2
UniProt ID: (Human) Q9Y2P8, (Mouse) Q9JJT0
Entrez Gene ID: (Human) 10171, (Mouse) 59028, (Rat) 309301
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support