Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: EREESEDELE EANGNNPIDI EVDQNKESKK EVPPTETVPQ VKKEKHSTQA KNRAKRKPPK GMFLSQEDVE AVSANATAAT TVLRQLDMEL VSVKRQIQNI KQTNSALKEK LDGGIEPYRL PEVIQKCNAR WTTEEQLLAV QAIRKYGRDF QAISDVIGNK SVVQVKNFFV NYRRRFNIDE VLQEWEAEHG KEETNGPSNQ KPVKSPDNSI KMPEEEDEAP VLDVRYASAS; Positive Samples: HeLa, Jurkat, Mouse testis, Mouse brain; Cellular Location: Nucleus
RCOR1 is a functional corepressor required for regulation of neural-specific gene expression. The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression. The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein CoREST; REST corepressor 1
Gene Aliases: 5730409O11; 6720480E22Rik; AU042633; COREST; D12Wsu95e; KIAA0071; mKIAA0071; RCOR; RCOR1; Rocr1
UniProt ID: (Human) Q9UKL0, (Mouse) Q8CFE3
Entrez Gene ID: (Human) 23186, (Mouse) 217864
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support