Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Mammalian odorant receptors require accessory proteins such as RTP1 and RTP2 for functional cell surface expression. Receptor expression-enhancing protein (REEP) family members are transmembrane proteins which interact with odorant receptor proteins and may enhance the odorant receptor responses to odorants. Recently studies have shown other chemosensory receptors such as bitter taste receptors are also influenced by RTP and REEP family members.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C2orf19; LOC682105; Receptor expression-enhancing protein 2
Gene Aliases: C5orf19; REEP2; SGC32445; SPG72; Yip2d
UniProt ID: (Human) Q9BRK0
Entrez Gene ID: (Human) 51308
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support