Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: RYERTDTGEI TSYITEDDVV YRPGDCVYIV CRRPNTPYFI CSIQDFKLVH NSQACCRSPT PALCDPPACS LPVASQPPQH LSEAGRGPVG SKRDHLLMNV KWYYRQSEV
Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arginine-glutamic acid dipeptide (RE) repeats; Arginine-glutamic acid dipeptide repeats protein; atrophin 2; atrophin-1 like protein; atrophin-1 related protein; Atrophin-1-like protein; Atrophin-1-related protein; FLJ38775
Gene Aliases: ARG; ARP; ATN1L; DNB1; KIAA0458; NEDBEH; RERE
UniProt ID: (Human) Q6P6B9
Entrez Gene ID: (Human) 473
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support