Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQAFLKGTS ISTKPPLTKD RGVAASAGSS GENKKAKPVP WVEKYRPKCV DEVAFQEEVV AVLKKSLEGA DLPNLLFYGP PGTGKTSTIL AAARELFGPE LFRLRVLELN ASDERGIQVV REKVKNFAQL TVSGSRSDGK PCPPFKIVIL DEADSMTSAA QAALRRTMEK ESKTTRFCLI CNYVSRIIEP LTSRCSKFRF KPLSDKIQQQ RLLDIAKKEN VKISDEGIAY LVKVSEGDLR KAITFLQSAT RLTGGKEITE KVITDIAGVI PAEKIDGVFA ACQSGSFDKL EAVVKDLIDE GHAATQLVNQ LHDVVVENNL SDKQKSIITE KLAEVDKCLA DGADEHLQLI SLCATVMQQL SQNC (1-363 aa encoded by BC017452)
Human Replication Factor C (RFC) is a hetero-pentameric protein consisting of the p140, p40, p38, p37 and p36 subunits. RFC functions in the loading of the DNA polymerase delta processivity factor (PCNA, Proliferating Cell nuclear Antigen) onto a primed DNA template during DNA synthesis and repair. RFC may also participate in the DNA damage checkpoint response with a PCNA-like hetero-trimer consisting of Hus 1, Rad 1 and Rad 9.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A1 37 kDa subunit; Activator 1 37 kDa subunit; Activator 1 subunit 4; replication factor C (activator 1) 4, 37kDa; Replication factor C 37 kDa subunit; Replication factor C subunit 4; RF-C 37 kDa subunit; RFC 37 kDa subunit; RFC p37
Gene Aliases: A1; RFC37; RFC4
UniProt ID: (Human) P35249
Entrez Gene ID: (Human) 5984
Molecular Function:
DNA-directed DNA polymerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support