Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Recommended positive controls: U87-MG, mouse liver.
Predicted reactivity: Human (86%), Mouse (86%), Rat (87%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Has E3 ubiquitin protein ligase activity. Regulates the levels of CASP8 and CASP10 by targeting them for proteasomal degradation. Has anti-apoptotic activity. May bind phosphatidylinositol phosphates.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CARP-2; Caspase regulator CARP2; Caspases-8 and -10-associated RING finger protein 2; E3 ubiquitin-protein ligase rififylin; Fring; FYVE-RING finger protein SAKURA; rififylin; ring finger and FYVE-like domain containing 1; RING finger and FYVE-like domain-containing protein 1; RING finger protein 189; RING finger protein 34-like; RING-type E3 ubiquitin transferase rififylin
Gene Aliases: 1700051E09Rik; 4930516L10Rik; BG080975; CARP-2; CARP2; FRING; RFFL; RIFIFYLIN; RNF189; RNF34L
UniProt ID: (Human) Q8WZ73, (Mouse) Q6ZQM0
Entrez Gene ID: (Human) 117584, (Mouse) 67338
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support