Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: LGSFMSTAEQ VRLPDDCTVG YIVEGLLGAR LLHSPLFHSH LENLQRLPPD TLLQQVTLSH GGPENPQNVV NVAGGFSLHQ DPTRFKSIHC LLYPDTDWCP RQKQGAPTS
Glycosyltransferase that initiates the elongation of O- linked fucose residues attached to EGF-like repeats in the extracellular domain of Notch molecules. May be involved in limb formation and in neurogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,3-N-acetylglucosaminyltransferase radical fringe; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; radical fringe homolog
Gene Aliases: RFNG
UniProt ID: (Human) Q9Y644
Entrez Gene ID: (Human) 5986
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support