Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPGMFFSANP KELKGTTHSL LDDKMQKRRP KTFGMDMKAY LRSMIPHLES GMKSSKSKDV LSAAEVMQWS QSLEKLLANQ TGQNVFGSFL KSEFSEENIE FWLACEDYKK TESDLLPCKA EEIYKAFVHS DAAKQINIDF RTRESTAKKI KAPTPTCFDE AQKVIYTLME KDSYPRFLKS DIYLNLLNDL QANSLK
This gene encodes a member of the regulator of G-protein signaling family. This protein is located on the cytosolic side of the plasma membrane and contains a conserved, 120 amino acid motif called the RGS domain. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell activation protein BL34; Early response protein 1R20; epididymis secretory protein Li 87; immediate-early response 1, B-cell specific; Regulator of G-protein signaling 1; regulator of G-protein signalling 1; RGS1
Gene Aliases: 1R20; BL34; HEL-S-87; IER1; IR20; RGS1
UniProt ID: (Human) Q08116
Entrez Gene ID: (Human) 5996
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support