Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWNTLTRSLS DHPVGKDPQA MRTGQRQNKG MRTRLGCLSH KSDSCSDFTA ILPDKPNRAL KRLSTEEATR WADSFDVLLS HKYGVAAFRA FLKTEFSEEN LEFWLACEEF KKTRSTAKLV SKAHRIFEEF VDVQAPREVN IDFQTREATR KNLQEPSLTC FDQAQGKVHS LMEKDSYPRF LRSKMYLDLL SQSQRRLS
A novel human RGS8 gene (homologue of the rat RGS8 gene) is expressed in prostate tissues and represents a positional candidate for the HPC1 locus, which is involved in hereditary prostate cancer and hyperparathyroidism-jaw tumor syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Regulator of G-protein signaling 8; regulator of G-protein signalling 8; RGS8
Gene Aliases: RGS8
UniProt ID: (Human) P57771
Entrez Gene ID: (Human) 85397
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support