Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
RHBG encodes one of two non-erythroid members of the Rhesus (Rh) protein family. Non-erythroid Rh protein family members are mainly expressed in the kidney and belong to the methylammonium-ammonium permease/ammonia transporters superfamily. All Rh family proteins are predicted to be transmembrane proteins with 12 membrane spanning domains and intracytoplasmic N- and C-termini.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ammonium transporter Rh type B; Rh family type B glycoprotein; Rh family, B glycoprotein; Rh type B glycoprotein; Rhesus blood group family type B glycoprotein; Rhesus blood group, B glycoprotein
Gene Aliases: RHBG; SLC42A2
UniProt ID: (Human) Q9H310, (Mouse) Q8BUX5
Entrez Gene ID: (Human) 57127, (Mouse) 58176
Molecular Function: primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support