Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: LPFWGQPSDE NCFEDAVYWE MPEGNSTVYI PEDPTFKPSG PSVPSVPMVS PLPMASSVPL VP
RHCG (Rhesus blood group family type C glycoprotein), also known as RHGK (Rh glycoprotein kidney), PDRC2 (tumor-related protein DRC2) or SLC42A3, is a multi-pass membrane protein with three potential N-glycosylation sites that belongs to the Rh subfamily of the ammonium transporter family. Expressed in a wide variety of tissues with predominant expression in adult and fetal kidney, RHCg localizes to the apical and/or basolateral cell membrane (depending on the species) and is believed to function as a non-erythroid, bidirectional, electroneutral ammonium transporter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ammonium transporter Rh type C; Rh family; Rh family type C glycoprotein; Rh family, C glycoprotein; Rh glycoprotein kidney; Rh type C glycoprotein; Rhesus blood group family type C glycoprotein; Rhesus blood group, C glycoprotein; Rhesus blood group-associated C glycoprotein; Tumor-related protein DRC2
Gene Aliases: C15orf6; CDRC2; PDRC2; RHCG; RHGK; SLC42A3
UniProt ID: (Human) Q9UBD6, (Rat) Q7TNN9
Entrez Gene ID: (Human) 51458, (Rat) 293048
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support