Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
May have a role in sperm development and fertilization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp434D1026; DKFZp781N1478; FLJ12870; RAP1 interacting factor homolog; Rap1-interacting factor 1 homolog; Telomere-associated protein RIF1
Gene Aliases: RIF1
UniProt ID: (Human) Q5UIP0
Entrez Gene ID: (Human) 55183
Molecular Function:
DNA metabolism protein
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support