Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalyzes the synthesis of N-acetyl-L-aspartyl-L-glutamate (NAAG) and N-acetyl-L-aspartyl-L-glutamyl-L-glutamate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 80, member A; N-acetylaspartyl-glutamate synthetase A; N-acetylaspartylglutamate synthase A; N-acetylaspartylglutamate synthetase II; N-acetylaspartylglutamylglutamate synthase A; NAAG synthetase A; ribosomal modification protein rimK-like family member A; Ribosomal protein S6 modification-like protein A; RP11-157D18.1
Gene Aliases: B930030J24; FAM80A; NAAGS; NAAGS-II; Rimk; RIMKLA
UniProt ID: (Human) Q8IXN7, (Mouse) Q6PFX8
Entrez Gene ID: (Human) 284716, (Mouse) 194237
Molecular Function: ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support