Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MCSSVAAKL WFLTDRRIRE DYPQKEILRA LKAKCCEEEL DFRAVVMDEV VLTIEQGNLG LRINGELITA YPQVVVVRVP TPWVQSDSDI TVLRHLEKMG CRLMNRPQAI LNCVNKFWTF QELAGHGVPL PDTFSYGGHE NFAKMIDEAE VLEFPMVVKN TRGHRGKAVF LARDKHHLAD LSHLIRHEAP YLFQKYVKES HGRDVRVIVV GGRVVGTMLR CSTDGRMQSN CSLGGVGMMC SLSEQGKQLA IQVSNILGMD VCGIDLLMKD DGSFCVCEAN ANPFQEPKKQ IQTNKKIPRE KTFLLPPS (1-307 aa encoded by BC033793)
Catalyzes the synthesis of beta-citryl-L-glutamate and N-acetyl-L-aspartyl-L-glutamate. Beta-citryl-L-glutamate is synthesized more efficiently than N-acetyl-L-aspartyl-L-glutamate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-citryl-glutamate synthase B; Beta-citrylglutamate synthase B; family with sequence similarity 80, member B; N-acetyl-aspartyl-glutamate synthetase B; N-acetyl-aspartylglutamate synthetase B; N-acetylaspartyl-glutamate synthetase B; NAAG synthetase B; ribosomal modification protein rimK-like family member B; Ribosomal protein S6 modification-like protein B
Gene Aliases: 4931417E21Rik; 4933426K21Rik; AA097693; AI325948; AU015629; FAM80B; KIAA1238; NAAGS; NAAGS-I; RIMKLB
UniProt ID: (Human) Q9ULI2, (Mouse) Q80WS1
Entrez Gene ID: (Human) 57494, (Mouse) 108653
Molecular Function: ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support