Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Has E3 ubiquitin protein ligase activity. Regulates the levels of CASP8 and CASP10 by targeting them for proteasomal degradation. Has anti-apoptotic activity. May bind phosphatidylinositol phosphates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CARP-2; Caspase regulator CARP2; Caspases-8 and -10-associated RING finger protein 2; E3 ubiquitin-protein ligase rififylin; Fring; FYVE-RING finger protein SAKURA; rififylin; ring finger and FYVE like domain containing protein; ring finger and FYVE-like domain containing 1; RING finger and FYVE-like domain-containing protein 1; RING finger protein 189; RING finger protein 34-like; RING finger protein SAKURA; RING-type E3 ubiquitin transferase rififylin
Gene Aliases: 1700051E09Rik; 4930516L10Rik; BG080975; CARP-2; CARP2; FRING; RFFL; RIFIFYLIN; RNF189; RNF34L
UniProt ID: (Human) Q8WZ73, (Mouse) Q6ZQM0, (Rat) Q8CIN9
Entrez Gene ID: (Human) 117584, (Mouse) 67338, (Rat) 282844
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support