Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
RIOK2 belongs to the protein kinase superfamily; RIO-type Ser/Thr kinase family. Serine/threonine protein kinases, such as RIOK2, phosphorylate the OH group of serine or threonine (which have similar sidechains). Activity of these protein kinases can be regulated by specific events (e.g. DNA damage), as well as numerous chemical signals, including cAMP/cGMP, Diacylglycerol, and Ca2+/calmodulin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RIO kinase 2; Serine/threonine-protein kinase RIO2
Gene Aliases: 2010110K24Rik; 2410085M17Rik; RIO2; RIOK2
UniProt ID: (Human) Q9BVS4, (Mouse) Q9CQS5
Entrez Gene ID: (Human) 55781, (Mouse) 67045
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support