Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
RIPK5 is a serine/threonine kinase that is involved in cell activation, proliferation, differentiation, and apoptosis. This gene encodes a dual serine/threonine and tyrosine protein kinase which is expressed in multiple tissues. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dual serine/threonine and tyrosine protein kinase; Dusty PK; Dusty protein kinase; receptor interacting protein kinase 5; Receptor-interacting serine/threonine-protein kinase 5; RIP-homologous kinase; SgK496; Sugen kinase 496
Gene Aliases: A930019K20Rik; C430014H23Rik; C820013G01; CAKUT1; DSTYK; DustyPK; HDCMD38P; KIAA0472; RIP5; RIPK5; SGK496
UniProt ID: (Bovine) Q4TVR5, (Human) Q6XUX3, (Rat) Q6XUX2, (Mouse) Q6XUX1
Entrez Gene ID: (Bovine) 534684, (Human) 25778, (Rat) 304791, (Mouse) 213452
Molecular Function:
non-receptor serine/threonine protein kinase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support