Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MSALTPPTDM PTPTTDKITQ AAMETIYLCK FRVSMDGEWL CLRELDDISL TPDPEPTHED PNYLM; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell projection, Cytoplasm, Endomembrane system, Perikaryon, filopodium, growth cone, invadopodium, lamellipodium
Implicated in the formation of a single axon by developing neurons. May inhibit the formation of additional axons by inhibition of PI3K in minor neuronal processes.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein RUFY3; rap2 interacting protein x; Rap2-interacting protein x; RIPx; RUN and FYVE domain-containing protein 3; Singar; Singar1; singar2; Single axon-regulated protein; Single axon-regulated protein 1
Gene Aliases: 2810428M05Rik; 6330416M07Rik; AW455998; AW538594; D5Bwg0860e; Ripx; Rpipx; Rufy3; Singar1
UniProt ID: (Mouse) Q9D394, (Rat) Q5FVJ0
Entrez Gene ID: (Mouse) 52822, (Rat) 360921
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support