Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: KESRAKKFQR QHMDSDSSPS SSSTYCNQMM RRRNMTQGRC KPVNTFVHEP LVDVQNVCFQ EKVTCKNGQG NCYKSNSSMH ITDCRLTNGS RYPNCAYRTS
Endonuclease that catalyzes the cleavage of RNA on the 3' side of pyrimidine nucleotides. Acts on single stranded and double stranded RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HP-RNase; Rib 1; RIB-1; Ribonuclease 1; Ribonuclease A; ribonuclease A C1; Ribonuclease pancreatic; ribonuclease, RNase A family, 1 (pancreatic); RNase 1; RNase A; RNase UpI-1; RNAseA
Gene Aliases: RAC1; RIB1; RNASE1; RNS1
UniProt ID: (Human) P07998
Entrez Gene ID: (Human) 6035
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support