Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
RNASE8 has a low ribonuclease activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ribonuclease 8; ribonuclease, RNase A family, 8; RNase 8; RNase 8, placental
Gene Aliases: RAE2; RNASE8
UniProt ID: (Human) Q8TDE3
Entrez Gene ID: (Human) 122665
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support