Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MESGDEAAI ERHRVHLRSA TLRDAVPATL HLLPCEVAVD GPAPVGRFFT PAIRQGPEGL EVSFRGRCLR GEEVAVPPGL VGYVMVTEEK KVSMGKPDPL RDSGTDDQEE EPLERDFDRF IGATANFSRF TLWGLETIPG PDAKVRGALT WPSLAAAIHA QVPED (1-164 aa encoded by BC023588)
RNH2C encodes a ribonuclease H subunit that can cleave ribonucleotides from RNA:DNA duplexes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGS3; Aicardi-Goutieres syndrome 3 protein; Ribonuclease H2 subunit C; ribonuclease H2, subunit C; Ribonuclease HI subunit C; RNase H1 small subunit; RNase H2 subunit C
Gene Aliases: AGS3; AYP1; RNASEH2C
UniProt ID: (Human) Q8TDP1
Entrez Gene ID: (Human) 84153
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support