Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Positive Samples: HeLa, HepG2, K562, mouse testis, rat liver
Immunogen sequence: TGPPPADKEK IQALPTVPVT EEHVGSGLEC PVCKDDYALG ERVRQLPCNH LFHDGCIVPW LEQHDSCPVC RKSLTGQNTA TNPPGLTGVS FSSSSSSSSS S
The specific function of RNF126 is not yet known. The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RNF126; RING finger protein 126
Gene Aliases: 2610010O19Rik; RNF126
UniProt ID: (Human) Q9BV68, (Mouse) Q91YL2
Entrez Gene ID: (Human) 55658, (Mouse) 70294, (Rat) 314613
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support