Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAEASPHPG RYFCHCCSVE IVPRLPDYIC PRCESGFIEE LPEETRSTEN GSAPSTAPTD QSRPPLEHVD QHLFTLPQGY GQFAFGIFDD SFEIPTFPPG AQADDGRDPE SRRERDHPSR HRYGARQPRA RLTTRRATGR HEGVPTLEGI IQQLVNGIIT PATIPSLGPW GVLHSNPMDY AWGANGLDAI ITQLLNQFEN TGPPPADKEK IQALPTVPVT EEHVGSGLEC PVCKDDYALG ERVRQLPCNH LFHDGCIVPW LEQHDSCPVC RKSLTGQNTA TNPPGLTGVS FSSSSSSSSS SSPSNENATW SPLGRPQPPR PLSNLTL (1-326 aa encoded by BC001442)
The specific function of RNF126 is not yet known. The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RNF126; RING finger protein 126
Gene Aliases: 2610010O19Rik; RNF126
UniProt ID: (Human) Q9BV68, (Mouse) Q91YL2
Entrez Gene ID: (Human) 55658, (Mouse) 70294, (Rat) 314613
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support