Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKRSKELITK NHSQEETSIL RCWKCRKCIA SSGCFMEYLE NQVIKDKDDS VDAQNICHVW HMNVEALPEW ISCLIQKAQW TVGKLNCPFC GARLGGFNFV
E3 ubiquitin-protein ligase which promotes polyubiquitination and degradation by the proteasome pathway of ZIC2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RNF180; RING finger protein 180; RING-type E3 ubiquitin transferase RNF180
Gene Aliases: RINES; RNF180
UniProt ID: (Human) Q86T96
Entrez Gene ID: (Human) 285671
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support