Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: YWSQFDENI RIILKRYDLE QGLGDLLTER KALVVPEPEP DSDSNQERKD DRERGEGQEP AFSFLATLAS SSSEEMESQL QERVESSRRA VSQIVTVYDK LQEKVELLSR KLNSGDNLIV EEAVQELNSF LAQENMRLQE LTDLLQEKHR TMSQEFSKLQ SKVETAESRV SVLESMIDDL QWDIDKIRKR EQRLNRHLAE VLERVNSKGY KVYGAGSSLY GGTITINARK FEEMNAELEE NKELAQNRLC ELEKLRQDFE EVTTQNEKLK VELRSAVEQV VKETPEYRCM QSQFSVLYNE SLQLKAHLDE ARTLLHGTRG THQHQVELIE RDEVSLHKK (96-433 aa encoded by BC110585)
RNF20 is an E3 ubiquitin ligase that regulates chromosome structure by monoubiquitinating histone H2B. RNF20 selectively suppresses the expression of some genes such as several protooncogenes and growth-related genes, including many genes that are induced by epidermal growth factor by interfering with chromatin recruitment of transcription elongation factor SII (TFIIS). In contrast, RNF20 also positively regulates the p53 tumor suppressor as well as numerous histone H2A and H2B genes. Together with the deubiquitinase USP44, RNF20 regulates stem cell differentiation by modulating H2B monoubiquitination.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRE1 E3 ubiquitin ligase homolog; BRE1-A; E3 ubiquitin-protein ligase BRE1A; FLJ11189; FLJ20382; HNF3B; homolog of S. cerevisiae BRE1; KIAA2779; MGC129667; MGC129668; RING finger protein 20; RING-type E3 ubiquitin transferase BRE1A; TCF3B
Gene Aliases: 4833430L21Rik; AW540162; BRE1; BRE1A; C79397; hBRE1; Kiaa4116; mKIAA4116; RNF20
UniProt ID: (Human) Q5VTR2, (Mouse) Q5DTM8
Entrez Gene ID: (Human) 56254, (Mouse) 109331, (Rat) 313216
Molecular Function:
immunoglobulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support