Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGYDVTRFQ GDVDEDLICP ICSGVLEEPV QAPHCEHAFC NACITQWFSQ QQTCPVDRSV VTVAHLRPVP RIMRNMLSKL QIACDNAVFG CSAVVRLDNL MSHLSDCEHN PKRPVTCEQG CGLEMPKDEL PNHNCIKHLR SVVQQQQTRI AELEKTSAEH KHQLAEQKRD IQLLKAYMRA IRSVNPNLQN LEETIEYNEI LEWVNSLQPA RVTRWGGMIS TPDAVLQAVI KRSLVESGCP ASIVNELIEN AHERSWPQGL ATLETRQMNR RYYENYVAKR IPGKQAVVVM ACENQHMGDD MVQEPGLVMI FAHGVEEI (1-317 aa encoded by BC032637)
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. Nrdp1, also known as RNF41 (RING finger protein 41), SBBI03 or FLRF, is a 317 amino acid protein that contains one RING-type zinc finger and one SIAH-type zinc finger. Expressed in testis, ovary and prostate, Nrdp1 functions as an E3 ubiquitin-protein ligase that, characteristic of E3 ligase proteins, accepts ubiquitin (in the form of a thioester) from an E2 ubiquitin-conjugating enzyme and transfers that ubiquitin residue to substrates targeted for degradation. Specifically, Nrdp1 interacts with ErbB-3 and UBPY, thereby targeting them for proteasomal degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase NRDP1; fetal liver ring finger; neuregulin receptor degradation protein-1; RING finger protein 41; ring finger protein 41, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase NRDP1
Gene Aliases: 2210404G21Rik; 4930511A05Rik; 4933415P08Rik; D10Ertd722e; FLRF; NRDP1; RNF41; SBBI03
UniProt ID: (Human) Q9H4P4, (Mouse) Q9CRK6
Entrez Gene ID: (Human) 10193, (Mouse) 67588, (Rat) 362814
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support