Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: TQCFQLSNMF NPQTEEEVGW DTEIKDDVIE ECNKHGGVIH IYVDKNSAQG
The protein encoded by this gene is an RNA binding protein and possible splicing factor. The encoded protein is found in the nucleus, where it colocalizes with core spliceosomal proteins. Studies of a mouse protein with high sequence similarity to this protein suggest that this protein may act as a transcriptional coactivator for JUN/AP-1 and estrogen receptors. Multiple transcript variants encoding different isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAPER alpha; CAPERalpha; Coactivator of activating protein 1 and estrogen receptors; coactivator of activating protein-1 and estrogen receptors; Coactivator of AP-1 and ERs; functional spliceosome-associated protein 59; Hepatocellular carcinoma protein 1; RNA-binding motif protein 39; RNA-binding protein 39; RNA-binding region (RNP1, RRM) containing 2; RNA-binding region-containing protein 2; Splicing factor HCC1; transcription coactivator CAPER
Gene Aliases: 1500012C14Rik; 2310040E03Rik; B330012G18Rik; C79248; CAPER; CAPERalpha; FSAP59; HCC1; R75070; RBM39; RNPC2
UniProt ID: (Human) Q14498, (Mouse) Q8VH51
Entrez Gene ID: (Human) 9584, (Mouse) 170791
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support