Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Recommended positive controls: 293T, A431, HeLa, HepG2.
Predicted reactivity: Mouse (100%), Rat (98%), Chimpanzee (98%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. This protein contains many serine residues. Two splice variants have been found for this gene; both variants encode the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA binding protein S1, serine-rich domain; RNA binding protein with serine-rich domain; RNA-binding protein S1, serine-rich domain; RNA-binding protein with serine-rich domain 1; SR protein; SR-related protein LDC2
Gene Aliases: E5.1; LDC2; RNPS1
UniProt ID: (Human) Q15287
Entrez Gene ID: (Human) 10921
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support