Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MESGDEAAIE RHRVHLRSAT LRDAVPATLH LLPCEVAVDG PAPVGRFFTP AIRQGPEGLE VSFRGRCLRG EEVAVPPGLV GYVMVTEEKK VSMGKPDPLR DSGTDDQEEE PLERDFDRFI GATANFSRFT LWGLETIPGP DAKVRGALTW PSLAAAIHAQ VPED; Positive Samples: 293T, HeLa; Cellular Location: Nucleus
RNH2C encodes a ribonuclease H subunit that can cleave ribonucleotides from RNA:DNA duplexes.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGS3; Aicardi-Goutieres syndrome 3 protein; Ribonuclease H2 subunit C; ribonuclease H2, subunit C; Ribonuclease HI subunit C; RNase H1 small subunit; RNase H2 subunit C
Gene Aliases: AGS3; AYP1; RNASEH2C
UniProt ID: (Human) Q8TDP1
Entrez Gene ID: (Human) 84153
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support