Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ36151; HsROD; Kinetochore-associated protein 1; Rough deal homolog; Rough Deal homolog, centromere/kinetochore protein
Gene Aliases: KIAA0166; KNTC1; ROD
UniProt ID: (Human) P50748
Entrez Gene ID: (Human) 9735
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support