Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Immunogen sequence: ASFSDSEDE SCVPLLRKES IQLRDLDSAL LAEVKDVLIP HERVVTHSDR VIGKGHFGVV YHGEYIDQAQ NRIQCAIKSL SRITEMQQVE AFLREGLLMR GLNHPNVLAL IGIMLPPEGL PHVLLPYMCH GDLLQFIRSP QRNPTVKDLI SFGLQVARGM EYLAEQKFVH RDLAARNCML DESFTVKVAD FGLARDILDR EYYSVQQHRH ARLPVKWMAL ESLQTYRFTT KSDVWSFGVL LWELLTRGAP PYRHIDPFDL THFLAQGRRL PQPEYCPDSL YQVMQQCWEA DPAVRP (1038-1332 aa encoded by BQ936677 NM_002447)
MST1R (RON) is a receptor tyrosine kinase involved in cell proliferation and motility. The protein is a membrane-spanning, disulfide-linked heterodimer, which results from cleavage of a glycosylated precursor into 35-kD (alpha) and 150-kD (beta) subunits. Ligand binding results in tyrosine phosphorylation of the beta chain. In knockout studies, MST1R/RON (-/-) mice failed to survive past the periimplantation period. The MST1R/RON gene has been mapped to 3p21, a region of frequent deletion or mutation in small cell lung and renal carcinoma, and has been implicated in the progression of several epithelial cancers. MST1R/Ron expression has been documented in many normal human tissues. ESTs have been isolated from several tissue libraries, including normal colon, mouth, prostate, and testis and cancerous colon, prostate, stomach, and uteru.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-met-related tyrosine kinase; CD136; CDw136; friend virus susceptibility 2; Macrophage-stimulating protein receptor; MSP receptor; MST1R variant RON30; MST1R variant RON62; p185-Ron; protein tyrosine kinase 8; Protein-tyrosine kinase 8; PTK8 protein tyrosine kinase 8; receptor protein tyrosine kinase, c-met-related; RON variant E2E3; Stem cell-derived tyrosine kinase
Gene Aliases: CD136; CDw136; Fv-2; Fv2; MST1R; PTK8; RON; STK
UniProt ID: (Human) Q04912, (Mouse) Q62190
Entrez Gene ID: (Human) 4486, (Rat) 300999, (Mouse) 19882
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support