Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGSKLTCCLG PSGGLNCDCC RPDVGPCHEC EIPETVAATA PASTTAKPAK LDLKAKKAQL MQYLSLPKTP KMLKMSKGLD ARSKRWLKII WRRHGIWPLE NIGPTEDVQA SAHGGVEENM TSDIEIPEAK HDHRPTEDVQ VSAHGGVEEN ITSDIEISEA KHDHHLVEDL SESLSVCLED FMTSDLSESL SVSLEDFMTS GLSESLSVSL EDLMTPEMAK ERYEDYLCWV KMARSRLNEP ISSQVLGLLR LPTFLY
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein C22orf42
Gene Aliases: C22orf42; dJ90G24.6
UniProt ID: (Human) Q6IC83
Entrez Gene ID: (Human) 150297
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support