Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASLYVGDLH PEVTEAMLYE KFSPAGPILS IRICRDKITR RSLGYAYVNY QQPVDAKRAL ETLNFDVIKG RPVRIMWSQR DPSLRKSGVG NVFIKNLGKT IDNKALYNIF SAFGNILSCK VACDEKGPKG YGFVHFQKQE SAERAIDVMN GMFLNYRKIF VGRFKSHKER EAERGAWARQ STSADVKDFE EDTDEEATLR
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: poly(A) binding protein, cytoplasmic 1-like 2A; Polyadenylate-binding protein 1-like 2; RNA binding motif protein 32A; RNA-binding motif protein 32; RNA-binding protein 32
Gene Aliases: PABPC1L2; PABPC1L2A; PABPC1L2B; RBM32A; RBM32B
UniProt ID: (Human) Q5JQF8
Entrez Gene ID: (Human) 340529
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support