Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKRVNSCVKS DEHVLEELET EGERQLKSLL QHQLDTSVSI EECMSKKESF APGTMYKPFG KEAAGTMTLS QFQTLHEKDQ ETASLRELGL NETEILIWKS HVSGEKKTKL RATPEAIQNR LQDIEERISE RQRILCLPQR FAKSKQLTRR EMEIEKSLFQ GADRHSFLKA LYYQDEPQKK NKGDPMNNLE SFYQEMIMKK RLEEFQLMRG EPFASHSLVS ATSVGDSGTA ESPSLLQDKG KQAAQGKGPS LHVANVIDFS PEQCWTGPKK LTQPIEFVPE DEIQRNRLSE EEIRKIPMFS SYNPGEPNKV LYLKNLSPRV TERDLVSLFA RFQEKKGPPI QFRMMTGRMR GQAFITFPNK EIAWQALHLV NGYKLYGKIL VIEFGKNKKQ RSNLQATSLI SCATGSTTEI SGS
May bind RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA-binding motif protein 41; RNA-binding protein 41
Gene Aliases: RBM41
UniProt ID: (Human) Q96IZ5
Entrez Gene ID: (Human) 55285
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support