Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGCFFSKRR KADKESRPEN EEERPKQYSW DQREKVDPKD YMFSGLKDET VGRLPGTVAG QQFLIQDCEN CNIYIFDHSA TVTIDDCTNC IIFLGPVKGS VFFRNCRDCK CTLACQQFRV RDCRKLEVFL CCATQPIIES SSNIKFGCFQ WYYPELAFQF KDAGLSIFNN TWSNIHDFTP VSGELNWSLL PEDAVVQDYV PIPTTEELKA VRVSTEANRS IVPISRGQRQ KSSDESCLVV LFAGDYTIAN ARKLIDEMVG KGFFLVQTKE VSMKAEDAQR VFREKAPDFL PLLNKGPVIA LEFNGDGAVE VCQLIVNEIF NGTKMFVSES KETASGDVDS FYNFADIQMG I (1-350 aa encoded by BC043348)
The RP2 locus has been implicated as one cause of X-linked retinitis pigmentosa. The predicted gene product shows homology with human cofactor C, a protein involved in the ultimate step of beta-tubulin folding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIAA0215; Protein XRP2
Gene Aliases: AI662636; DELXp11.3; NM23-H10; NME10; RGD1565124; RP2; Rp2h; TBCCD2; XRP2
UniProt ID: (Human) O75695, (Mouse) Q9EPK2
Entrez Gene ID: (Human) 6102, (Rat) 367714, (Mouse) 19889
Molecular Function:
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support